COMH42117533 Combi-Blocks, Inc.

2-(2-Hydroxybenzoyl)hydrazinecarboximidamide

  • MDL number MFCD20713541
  • Cas Number : 6005-61-4
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5283

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5283
Availability details for
Quantity
  • Availability
  • Add to Cart