COMH42117538 Combi-Blocks, Inc.

Methyl 5-methyl-2-phenyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD31619048
  • Cas Number : 856864-54-5
  • Molecular Weight : 268.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5314

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5314
Availability details for
Quantity
  • Availability
  • Add to Cart