COMH4211753F Combi-Blocks, Inc.

Methyl 2-(acetoxy(4-chlorophenyl)methyl)acrylate

  • MDL number MFCD18378272
  • Cas Number : 131469-72-2
  • Molecular Weight : 268.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-[(acetyloxy)(4-chlorophenyl)methyl]acrylate
Inchi Code 1S/C13H13ClO4/c1-8(13(16)17-3)12(18-9(2)15)10-4-6-11(14)7-5-10/h4-7,12H,1H2,2-3H3
Inchi Key SUCGWHZXYIZOIK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5348

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5348
Availability details for
Quantity
  • Availability
  • Add to Cart