COMH42117544 Combi-Blocks, Inc.

N-(Acid-peg3)-n-bis(peg3-amine)

  • MDL number MFCD31580144
  • Cas Number : 2183440-35-7
  • Molecular Weight : 571.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5380
Availability details for
Quantity
  • Availability
  • Add to Cart