COMH42117548 Combi-Blocks, Inc.

3-Heptyl-1h-1,2,4-triazol-5-amine

  • MDL number MFCD31619041
  • Cas Number : 20586-95-2
  • Molecular Weight : 182.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5391
Availability details for
Quantity
  • Availability
  • Add to Cart