COMH4211754B Combi-Blocks, Inc.

4-[Benzyl(methyl)amino]-4-oxobutanoic acid

  • MDL number MFCD03193260
  • Cas Number : 69049-90-7
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5417
Availability details for
Quantity
  • Availability
  • Add to Cart