COMH42117553 Combi-Blocks, Inc.

3-(Pentylsulfinyl)-1h-1,2,4-triazol-5-amine

  • MDL number MFCD31619040
  • Cas Number : 2213552-11-3
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5459
Availability details for
Quantity
  • Availability
  • Add to Cart