COMH42117556 Combi-Blocks, Inc.

2-Chloro-n-[2-(3,4-dimethoxyphenyl)ethyl]acetamide

  • MDL number MFCD00187582
  • Cas Number : 14301-31-6
  • Molecular Weight : 257.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5479

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5479
Availability details for
Quantity
  • Availability
  • Add to Cart