COMH4211755C Combi-Blocks, Inc.

(2-Pyridyldithio)-peg2-t-butyl ester

  • MDL number MFCD31561120
  • Cas Number : 2144777-73-9
  • Molecular Weight : 359.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(2-(2-(pyridin-2-yldisulfaneyl)ethoxy)ethoxy)propanoate
Inchi Code 1S/C16H25NO4S2/c1-16(2,3)21-15(18)7-9-19-10-11-20-12-13-22-23-14-6-4-5-8-17-14/h4-6,8H,7,9-13H2,1-3H3
Inchi Key NFTUEHOZCVWJPT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5524
Availability details for
Quantity
  • Availability
  • Add to Cart