COMH4211755D Combi-Blocks, Inc.

3-(Butylsulfinyl)-1h-1,2,4-triazol-5-amine

  • MDL number MFCD31619039
  • Cas Number : 2215441-67-9
  • Molecular Weight : 188.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5528
Availability details for
Quantity
  • Availability
  • Add to Cart