COMH42117567 Combi-Blocks, Inc.

2-(2-Hydroxyethyl)-4-methylpentanoate sodium salt

  • MDL number MFCD31583447
  • Cas Number : 2270912-50-8
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name sodium 2-(2-hydroxyethyl)-4-methylpentanoate
Inchi Code 1S/C8H16O3.Na/c1-6(2)5-7(3-4-9)8(10)11;/h6-7,9H,3-5H2,1-2H3,(H,10,11);/q;+1/p-1
Inchi Key BEIQARRVEAETBY-UHFFFAOYSA-M

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5588

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5588
Availability details for
Quantity
  • Availability
  • Add to Cart