COMH4211756D Combi-Blocks, Inc.

3-(Difluoromethyl)-1-methyl-1h-pyrazole-5-carbonyl chloride

  • MDL number MFCD31557145
  • Cas Number : 2182594-97-2
  • Molecular Weight : 194.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(difluoromethyl)-1-methyl-1H-pyrazole-5-carbonyl chloride

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5609
Availability details for
Quantity
  • Availability
  • Add to Cart