COMH4211756F Combi-Blocks, Inc.

Isopropylhydrazine

  • MDL number MFCD03410233
  • Cas Number : 2257-52-5
  • Molecular Weight : 74.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-isopropylhydrazine
Inchi Code 1S/C3H10N2/c1-3(2)5-4/h3,5H,4H2,1-2H3
Inchi Key KJAQRHMKLVGSCG-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5618
Availability details for
Quantity
  • Availability
  • Add to Cart