COMH42117577 Combi-Blocks, Inc.

2-Methyl-3,4-dihydro-2h-thieno[2,3-e][1,2]thiazin-4-amine 1,1-dioxide hydrochloride

  • MDL number MFCD14279884
  • Cas Number : 1158261-16-5
  • Molecular Weight : 254.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5638
Availability details for
Quantity
  • Availability
  • Add to Cart