COMH4211757B Combi-Blocks, Inc.

4-(3,4-Dimethylphenyl)pyrimidin-2-amine

  • MDL number MFCD07381834
  • Cas Number : 1247384-22-0
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5651

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5651
Availability details for
Quantity
  • Availability
  • Add to Cart