COMH42117583 Combi-Blocks, Inc.

3-Amino-n-cyclopropyl-2-methylbenzamide

  • MDL number MFCD09046152
  • Cas Number : 926189-70-0
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5699

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5699
Availability details for
Quantity
  • Availability
  • Add to Cart