COMH42117587 Combi-Blocks, Inc.

2,2,3,3,5,5,6,6-Octafluoro-1,4-dioxane

  • MDL number MFCD27918494
  • Cas Number : 32981-22-9
  • Molecular Weight : 232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5740
Availability details for
Quantity
  • Availability
  • Add to Cart