COMH42117591 Combi-Blocks, Inc.

3-Anilino-1-prop-2-yn-1-yl-1,3-dihydro-2h-indol-2-one

  • MDL number MFCD12028078
  • Cas Number : 1169930-57-7
  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5797
Availability details for
Quantity
  • Availability
  • Add to Cart