COMH42117594 Combi-Blocks, Inc.

Ethyl (3-methyl-4-nitrophenoxy)acetate

  • MDL number MFCD03419384
  • Cas Number : 158425-72-0
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5805

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5805
Availability details for
Quantity
  • Availability
  • Add to Cart