COMH421175A2 Combi-Blocks, Inc.

N-(Thiophen-2-ylmethyl)cyclopentanamine hydrochloride

  • MDL number MFCD11983204
  • Cas Number : 1245568-74-4
  • Molecular Weight : 217.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5931
Availability details for
Quantity
  • Availability
  • Add to Cart