COMH421175A3 Combi-Blocks, Inc.

4-Methoxy-2,3,6-trimethyl-phenol

  • MDL number MFCD24713549
  • Cas Number : 53651-61-9
  • Molecular Weight : 166.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methoxy-2,3,6-trimethylphenol
Inchi Code 1S/C10H14O2/c1-6-5-9(12-4)7(2)8(3)10(6)11/h5,11H,1-4H3
Inchi Key SJIRRMZHJWPOCJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5946
Availability details for
Quantity
  • Availability
  • Add to Cart