COMH421175A7 Combi-Blocks, Inc.

N'-(1-Benzyl-3-bromo-1h-1,2,4-triazol-5-yl)formic hydrazide

  • MDL number MFCD31619036
  • Cas Number : 2279124-19-3
  • Molecular Weight : 296.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5955

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5955
Availability details for
Quantity
  • Availability
  • Add to Cart