COMH421175B4 Combi-Blocks, Inc.

(5-Hydroxy-1-benzofuran-3-yl)(3,4,5-trimethoxyphenyl)methanone

  • MDL number MFCD30188506
  • Cas Number : 1254339-38-2
  • Molecular Weight : 328.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6011
Availability details for
Quantity
  • Availability
  • Add to Cart