COMH421175B9 Combi-Blocks, Inc.

Ethyl 2-bromo-5-methyl[1,3]oxazolo[3,2-b][1,2,4]triazole-6-carboxylate

  • MDL number MFCD31583435
  • Cas Number : 2270908-40-0
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6030
Availability details for
Quantity
  • Availability
  • Add to Cart