COMH421175BF Combi-Blocks, Inc.

(2E)-2-(Isonicotinoylhydrazono)propanoic acid

  • MDL number MFCD00449361
  • Cas Number : 864363-27-9
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6046

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6046
Availability details for
Quantity
  • Availability
  • Add to Cart