COMH421175C2 Combi-Blocks, Inc.

4-Chloro-1-methyl-1h-pyrazole-3-carbonyl chloride

  • MDL number MFCD03421433
  • Cas Number : 1048340-09-5
  • Molecular Weight : 179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-1-methyl-1H-pyrazole-3-carbonyl chloride
Inchi Code 1S/C5H4Cl2N2O/c1-9-2-3(6)4(8-9)5(7)10/h2H,1H3
Inchi Key UGGVYWVYYZYMNZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6061
Availability details for
Quantity
  • Availability
  • Add to Cart