COMH421175C8 Combi-Blocks, Inc.

Cyclopentyl 4-aminobenzoate hydrochloride

  • MDL number MFCD08070526
  • Cas Number : 37005-75-7
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6086

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6086
Availability details for
Quantity
  • Availability
  • Add to Cart