COMH421175C9 Combi-Blocks, Inc.

[4-(Benzyloxy)-2-nitrophenyl]acetic acid

  • MDL number MFCD08142191
  • Cas Number : 6860-79-3
  • Molecular Weight : 287.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6088

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6088
Availability details for
Quantity
  • Availability
  • Add to Cart