COMH421175D7 Combi-Blocks, Inc.

N-Methyl-6-(pyrrolidin-3-ylmethyl)pyrimidin-4-amine hydrochloride

  • MDL number MFCD31537590
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6165
Availability details for
Quantity
  • Availability
  • Add to Cart