COMH421175D8 Combi-Blocks, Inc.

Fmoc-peg3-ala-ala-asn(trt)-pab-pnp

  • MDL number MFCD30458053
  • Cas Number : 2055042-82-3
  • Molecular Weight : 1212.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6166
Availability details for
Quantity
  • Availability
  • Add to Cart