COMH421175EB Combi-Blocks, Inc.

N-[(1,3-Dimethyl-1h-pyrazol-4-yl)methyl]-n-ethylamine

  • MDL number MFCD08457675
  • Cas Number : 957043-29-7
  • Molecular Weight : 153.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]-N-ethylamine
Inchi Code 1S/C8H15N3/c1-4-9-5-8-6-11(3)10-7(8)2/h6,9H,4-5H2,1-3H3
Inchi Key WIXGONMGSHBZFS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6268

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6268
Availability details for
Quantity
  • Availability
  • Add to Cart