COMH421175ED Combi-Blocks, Inc.

Methyl 2-amino-5,5-dimethyl-4,7-dihydro-5h-thieno[2,3-c]pyran-3-carboxylate

  • MDL number MFCD05999146
  • Cas Number : 713111-73-0
  • Molecular Weight : 241.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-5,5-dimethyl-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carboxylate
Inchi Code 1S/C11H15NO3S/c1-11(2)4-6-7(5-15-11)16-9(12)8(6)10(13)14-3/h4-5,12H2,1-3H3
Inchi Key SVGGVCOTTKLQGH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6271

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6271
Availability details for
Quantity
  • Availability
  • Add to Cart