COMH421175EE Combi-Blocks, Inc.

(S)-2-((S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)-5-ureidopentanoic acid

  • MDL number MFCD28139060
  • Cas Number : 159858-21-6
  • Molecular Weight : 496.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6275

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6275
Availability details for
Quantity
  • Availability
  • Add to Cart