COMH421175F9 Combi-Blocks, Inc.

4-(9,10-Dimethoxy-5,11-dioxoisoindolo[2,1-a]quinazolin-6(5h,6ah,11h)-yl)butanoic acid

  • MDL number MFCD28142497
  • Cas Number : 1630763-90-4
  • Molecular Weight : 396.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6350

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6350
Availability details for
Quantity
  • Availability
  • Add to Cart