COMH421175FC Combi-Blocks, Inc.

1-(2-Propen-1-yl)-4-piperidinecarboxylic acid ethyl ester

  • MDL number MFCD16151796
  • Cas Number : 177971-46-9
  • Molecular Weight : 197.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6360
Availability details for
Quantity
  • Availability
  • Add to Cart