COMH421175FF Combi-Blocks, Inc.

(2Z)-6-Hydroxy-2-(2,3,4-trimethoxybenzylidene)-1-benzofuran-3(2h)-one

  • MDL number MFCD04145698
  • Cas Number : 1638956-60-1
  • Molecular Weight : 328.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6378

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6378
Availability details for
Quantity
  • Availability
  • Add to Cart