COMH42117601 Combi-Blocks, Inc.

T-Boc-n-amido-peg2-nhs ester

  • MDL number MFCD31381627
  • Cas Number : 2183440-73-3
  • Molecular Weight : 374.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6380
Availability details for
Quantity
  • Availability
  • Add to Cart