COMH42117605 Combi-Blocks, Inc.

5-(Aminomethyl)pyrazolo[1,5-a]pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD31619055
  • Cas Number : 2279122-49-3
  • Molecular Weight : 227.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6390

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6390
Availability details for
Quantity
  • Availability
  • Add to Cart