COMH42117608 Combi-Blocks, Inc.

(3S,3Ar,5s,6as,7s)-1-methyl-2-oxooctahydro-3,5-methanocyclopenta[b]pyrrole-7-carboxylic acid

  • MDL number MFCD31614357
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6400
Availability details for
Quantity
  • Availability
  • Add to Cart