COMH4211760D Combi-Blocks, Inc.

2-(2-Methyl-4-oxo-1,4-dihydropyrimido[1,2-b]indazol-3-yl)acetic acid

  • MDL number MFCD28142526
  • Cas Number : 1630763-66-4
  • Molecular Weight : 257.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6435

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6435
Availability details for
Quantity
  • Availability
  • Add to Cart