COMH42117613 Combi-Blocks, Inc.

Dspe-peg4-acid

  • MDL number MFCD30828702
  • Cas Number : 2112738-58-4
  • Molecular Weight : 1010.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6454

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6454
Availability details for
Quantity
  • Availability
  • Add to Cart