COMH4211761B Combi-Blocks, Inc.

M-Peg6-2-methylacrylate

  • MDL number MFCD31561109
  • Cas Number : 90784-86-4
  • Molecular Weight : 364.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6488

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6488
Availability details for
Quantity
  • Availability
  • Add to Cart