COMH4211761F Combi-Blocks, Inc.

2-(Hydroxymethyl)-5-methoxy-1-methyl-4(1h)-pyridinone

  • MDL number MFCD22394214
  • Cas Number : 62885-45-4
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6521
Availability details for
Quantity
  • Availability
  • Add to Cart