COMH42117620 Combi-Blocks, Inc.

5-Bromo-2h-3,4'-bi-1,2,4-triazole

  • MDL number MFCD31619037
  • Cas Number : 233600-16-3
  • Molecular Weight : 215.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6523

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6523
Availability details for
Quantity
  • Availability
  • Add to Cart