COMH42117624 Combi-Blocks, Inc.

Sulfo dbco-maleimide

  • MDL number MFCD30458041
  • Cas Number : 2028281-86-7
  • Molecular Weight : 578.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6559

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6559
Availability details for
Quantity
  • Availability
  • Add to Cart