COMH42117626 Combi-Blocks, Inc.

4-(4-Ethoxyphenyl)pyrimidin-2-amine

  • MDL number MFCD07381849
  • Cas Number : 1251055-62-5
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6564

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6564
Availability details for
Quantity
  • Availability
  • Add to Cart