COMH42117628 Combi-Blocks, Inc.

2-(2-Methoxy-5-nitrophenoxy)ethanamine hydrochloride

  • MDL number MFCD31583453
  • Cas Number : 2270909-64-1
  • Molecular Weight : 248.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-methoxy-5-nitrophenoxy)ethan-1-amine hydrochloride
Inchi Code 1S/C9H12N2O4.ClH/c1-14-8-3-2-7(11(12)13)6-9(8)15-5-4-10;/h2-3,6H,4-5,10H2,1H3;1H
Inchi Key IEMPONYBZMAZSG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6576

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6576
Availability details for
Quantity
  • Availability
  • Add to Cart