COMH4211762A Combi-Blocks, Inc.

(4-Chlorophenyl)(5-hydroxy-1-benzofuran-3-yl)methanone

  • MDL number MFCD00430818
  • Cas Number : 17249-67-1
  • Molecular Weight : 272.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6588

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6588
Availability details for
Quantity
  • Availability
  • Add to Cart