COMH42117632 Combi-Blocks, Inc.

Biotin-peg3-nhboc

  • MDL number MFCD31561130
  • Cas Number : 418759-44-1
  • Molecular Weight : 518.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6629

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6629
Availability details for
Quantity
  • Availability
  • Add to Cart