COMH4211763E Combi-Blocks, Inc.

5-Chloro-1-methyl-1h-1,2,4-triazole-3-carboxamide

  • MDL number MFCD31557154
  • Cas Number : 2168214-27-3
  • Molecular Weight : 160.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6679

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6679
Availability details for
Quantity
  • Availability
  • Add to Cart